Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008430.3 | NP_001008430.2 | 5 - 6 | 402 | 81 | |
XM_012962908.2 | XP_012818362.1 | 5 - 6 | 586 | 81 | ||
XM_018093849.1 | XP_017949338.1 | 3 - 4 | 453 | 81 | ||
GenBank mRNA | AL779600.2 | - | 5 - 6 | 403 | 81 | |
BC080134.1 | - | 5 - 6 | 391 | 81 | ||
CF240668.1 | - | 5 - 6 | 391 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |