Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fmo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015783.1 | NP_001015783.1 | 4 - 5 | 641 | 62 | |
XM_012961025.2 | XP_012816479.1 | 5 - 6 | 848 | 62 | ||
GenBank mRNA | BC089725.1 | - | 4 - 5 | 641 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |