Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ggh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030427.1 | NP_001025598.1 | 5 - 6 | 629 | 115 | |
XM_012964805.2 | XP_012820259.1 | 5 - 6 | 713 | 115 | ||
GenBank mRNA | BC091047.1 | - | 5 - 6 | 629 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |