Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTTCAATAAAGATCATCCAAAG[C/T]CTCATTAGAAGGCTTGCACCAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607680 MIM: 612535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCHY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RCHY1 - ring finger and CHY zinc finger domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THAP6 - THAP domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317791.1 | Intron | NP_001304720.1 | ||||
NM_144721.5 | Intron | NP_653322.1 | ||||
XM_005262772.2 | Intron | XP_005262829.1 | ||||
XM_005262774.3 | Intron | XP_005262831.1 | ||||
XM_006714109.3 | Intron | XP_006714172.1 | ||||
XM_011531666.2 | Intron | XP_011529968.1 | ||||
XM_011531667.2 | Intron | XP_011529969.1 | ||||
XM_017007800.1 | Intron | XP_016863289.1 | ||||
XM_017007801.1 | Intron | XP_016863290.1 |