Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCAGATAAAGCTATGACATTAA[A/G]TTGTTAAAACTATCTTCTCCTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNJ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNJ - cyclin J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134375.1 | Intron | NP_001127847.1 | ||||
NM_001134376.1 | Intron | NP_001127848.1 | ||||
NM_019084.4 | Intron | NP_061957.2 | ||||
XM_005269921.3 | Intron | XP_005269978.1 | ||||
XM_011539885.2 | Intron | XP_011538187.1 | ||||
XM_017016354.1 | Intron | XP_016871843.1 | ||||
XM_017016355.1 | Intron | XP_016871844.1 | ||||
XM_017016356.1 | Intron | XP_016871845.1 |
ENTPD1-AS1 - ENTPD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3157 - microRNA 3157 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |