Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCGCTGTCCCCTGGCGAGGCGC[C/T]TTCGGGCGGGGCGGGGGGCGCGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612184 MIM: 606692 | ||||||||||||||||||||
Literature Links: |
CASKIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASKIN1 - CASK interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020764.3 | 4116 | Missense Mutation | AGC,GGC | S,G 1362 | NP_065815.1 |
TRAF7 - TNF receptor associated factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032271.2 | 4116 | Intron | NP_115647.2 | |||
XM_005255627.4 | 4116 | Intron | XP_005255684.1 | |||
XM_011522700.1 | 4116 | Intron | XP_011521002.1 |