Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCTCACTACTGCAGCCAACCCA[A/T]CTCCTGGAAGGGCGCTCCCGCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616332 MIM: 614917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMT1 - acidic residue methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286562.1 | 61 | Intron | NP_001273491.1 | |||
NM_024573.2 | 61 | Intron | NP_078849.1 |
RMND1 - required for meiotic nuclear division 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271937.1 | 61 | UTR 5 | NP_001258866.1 | |||
NM_017909.3 | 61 | UTR 5 | NP_060379.2 | |||
XM_005267040.3 | 61 | UTR 5 | XP_005267097.1 | |||
XM_017010988.1 | 61 | UTR 5 | XP_016866477.1 |