Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGTTATTGATCCCTGGGTTAA[C/T]TCAAACCATACTCGCTCACCCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601299 MIM: 608925 | ||||||||||||||||||||
Literature Links: |
BMPR1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BMPR1A - bone morphogenetic protein receptor type 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMRN2 - multimerin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024756.2 | 2941 | Silent Mutation | GAA,GAG | E,E 925 | NP_079032.2 | |
XM_005270153.1 | 2941 | Silent Mutation | GAA,GAG | E,E 864 | XP_005270210.1 | |
XM_006717970.3 | 2941 | Silent Mutation | GAA,GAG | E,E 864 | XP_006718033.1 |