Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACTGACTTGGTTTCTTTTGCAG[A/G]TAACCTTCTGTTCGGTGATGAAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604479 | ||||||||||||||||||||
Literature Links: |
SIRT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIRT1 - sirtuin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142498.1 | 484 | UTR 5 | NP_001135970.1 | |||
NM_001314049.1 | 484 | Intron | NP_001300978.1 | |||
NM_012238.4 | 484 | Missense Mutation | GAT,GGT | D,G 144 | NP_036370.2 |