Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGGAGCTGCAGCACTTGGGCGC[C/T]GCAGGGCACGAGGCAGTAGAGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611684 | ||||||||||||||||||||
Literature Links: |
FICD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FICD - FIC domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SART3 - squamous cell carcinoma antigen recognized by T-cells 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014706.3 | 2532 | Missense Mutation | CAG,CGG | Q,R 921 | NP_055521.1 | |
XM_005269241.4 | 2532 | Missense Mutation | CAG,CGG | Q,R 939 | XP_005269298.1 | |
XM_011539026.2 | 2532 | Missense Mutation | CAG,CGG | Q,R 633 | XP_011537328.1 |