Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCTTTTCCTCCATCTGTTGGC[A/G]ATAGTATTCCCGGTTGACAGCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615671 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SETD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SETD3 - SET domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032233.2 | 1486 | Missense Mutation | NP_115609.2 | |||
NM_199123.1 | 1486 | Intron | NP_954574.1 | |||
XM_005268127.3 | 1486 | Missense Mutation | XP_005268184.1 | |||
XM_011537231.2 | 1486 | Missense Mutation | XP_011535533.2 | |||
XM_011537232.2 | 1486 | Missense Mutation | XP_011535534.1 | |||
XM_011537233.2 | 1486 | Missense Mutation | XP_011535535.1 | |||
XM_011537234.1 | 1486 | Missense Mutation | XP_011535536.1 | |||
XM_011537235.1 | 1486 | Intron | XP_011535537.1 | |||
XM_017021699.1 | 1486 | Missense Mutation | XP_016877188.1 | |||
XM_017021700.1 | 1486 | Missense Mutation | XP_016877189.1 | |||
XM_017021701.1 | 1486 | Missense Mutation | XP_016877190.1 | |||
XM_017021702.1 | 1486 | Missense Mutation | XP_016877191.1 |