Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATTGTGAAGGACAGGGGCATCT[C/T]GGGGTTCTGGGGAGGCAAGGAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616318 MIM: 601795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDPD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDPD3 - glycerophosphodiester phosphodiesterase domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024307.2 | 659 | Missense Mutation | AAG,GAG | K,E 194 | NP_077283.2 |
LOC101928595 - uncharacterized LOC101928595 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK3 - mitogen-activated protein kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |