Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCCGATGTTCTCCACAGTCAGC[C/T]TGAACTCCTTGTACTCTGAGTAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607992 MIM: 606563 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUGP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUGP1 - SURP and G-patch domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172231.3 | Intron | NP_757386.2 | ||||
XM_005260002.2 | Intron | XP_005260059.1 | ||||
XM_011528155.2 | Intron | XP_011526457.1 |
TM6SF2 - transmembrane 6 superfamily member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |