Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGTTTATTTTCTTCCTAACTCA[C/T]TGCTGAATGGCCAAGCCAGGGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176265 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNC4 - potassium voltage-gated channel subfamily C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039574.2 | Intron | NP_001034663.1 | ||||
NM_004978.4 | Intron | NP_004969.2 | ||||
XM_006710625.3 | Intron | XP_006710688.1 | ||||
XM_006710627.3 | Intron | XP_006710690.1 | ||||
XM_011541401.2 | Intron | XP_011539703.1 | ||||
XM_011541402.1 | Intron | XP_011539704.1 | ||||
XM_011541403.1 | Intron | XP_011539705.1 | ||||
XM_011541404.1 | Intron | XP_011539706.1 |
KCNC4-AS1 - KCNC4 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |