Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCTCCATCTTTATTTCTGTCTC[C/T]ATCTTAATATCCTATACCTAGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TPM3 - tropomyosin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001043351.1 | 2869 | Intron | NP_001036816.1 | |||
NM_001043352.1 | 2869 | Intron | NP_001036817.1 | |||
NM_001043353.1 | 2869 | Intron | NP_001036818.1 | |||
NM_001278188.1 | 2869 | Intron | NP_001265117.1 | |||
NM_001278189.1 | 2869 | Intron | NP_001265118.1 | |||
NM_001278190.1 | 2869 | Intron | NP_001265119.1 | |||
NM_001278191.1 | 2869 | Intron | NP_001265120.1 | |||
NM_152263.3 | 2869 | UTR 3 | NP_689476.2 | |||
NM_153649.3 | 2869 | Intron | NP_705935.1 |