Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGACGTCTGGGACTCTGTGTCCA[C/T]TATCTCGTTTCCGGAAAAGGAGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147569 | ||||||||||||||||||||
Literature Links: |
IFNGR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFNGR2 - interferon gamma receptor 2 (interferon gamma transducer 1) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005534.3 | 1613 | Missense Mutation | ACT,ATT | T,I 322 | NP_005525.2 | |
XM_005260969.2 | 1613 | Missense Mutation | ACT,ATT | T,I 341 | XP_005261026.1 | |
XM_011529553.1 | 1613 | Missense Mutation | ACT,ATT | T,I 347 | XP_011527855.1 | |
XM_011529554.2 | 1613 | Missense Mutation | ACT,ATT | T,I 324 | XP_011527856.1 |
TMEM50B - transmembrane protein 50B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006134.6 | 1613 | Intron | NP_006125.2 | |||
XM_011529746.1 | 1613 | UTR 3 | XP_011528048.1 |