Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGGTGTCACAGCCGTCCGCGCC[A/G]GGCGACGTGCGGTTGCAGAGACGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNT7B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WNT7B - Wnt family member 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_058238.2 | 1271 | Silent Mutation | CCC,CCT | P,P 299 | NP_478679.1 | |
XM_011530366.1 | 1271 | Silent Mutation | CCC,CCT | P,P 303 | XP_011528668.1 |