Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTGGATTCATGCTGGTTATCGC[C/G]GTAATAAACTCAACTGCAGTGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IQCA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQCA1 - IQ motif containing with AAA domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270584.1 | 2381 | Silent Mutation | ACC,ACG | T,T 733 | NP_001257513.1 | |
NM_001270585.1 | 2381 | Silent Mutation | ACC,ACG | T,T 782 | NP_001257514.1 | |
NM_024726.4 | 2381 | Silent Mutation | ACC,ACG | T,T 774 | NP_079002.3 | |
XM_011511864.1 | 2381 | Silent Mutation | ACC,ACG | T,T 781 | XP_011510166.1 | |
XM_011511865.2 | 2381 | Silent Mutation | ACC,ACG | T,T 775 | XP_011510167.1 | |
XM_011511866.1 | 2381 | Silent Mutation | ACC,ACG | T,T 741 | XP_011510168.1 | |
XM_017004960.1 | 2381 | Silent Mutation | ACC,ACG | T,T 740 | XP_016860449.1 |