Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGATTTTCACTGCGTGTGCAGA[C/T]CTGCTTTTTTTCCTTATATCTGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IWS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IWS1 - IWS1, SUPT6H interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017969.2 | 2873 | Silent Mutation | AGA,AGG | R,R 803 | NP_060439.2 | |
XM_005263707.3 | 2873 | Silent Mutation | AGA,AGG | R,R 808 | XP_005263764.1 | |
XM_006712626.3 | 2873 | UTR 3 | XP_006712689.1 | |||
XM_011511450.1 | 2873 | Intron | XP_011509752.1 | |||
XM_011511452.1 | 2873 | Intron | XP_011509754.1 | |||
XM_017004467.1 | 2873 | UTR 3 | XP_016859956.1 | |||
XM_017004468.1 | 2873 | Silent Mutation | AGA,AGG | R,R 596 | XP_016859957.1 |