Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCTGTTTTCATCTTTTTTCCTA[A/T]CAGCCTGGAGTTGGTCTGGGGAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609316 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM31 - tripartite motif containing 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007028.4 | 1009 | Missense Mutation | GAT,GTT | D,V 300 | NP_008959.3 | |
XM_011514264.1 | 1009 | Missense Mutation | GAT,GTT | D,V 299 | XP_011512566.1 | |
XM_011514265.1 | 1009 | Missense Mutation | GAT,GTT | D,V 300 | XP_011512567.1 | |
XM_011514266.1 | 1009 | Intron | XP_011512568.1 |
TRIM31-AS1 - TRIM31 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |