Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGACAGTTGTTCAAGAAAGAGA[G/T]CCTTTGGTGTGCTAAAGACTCTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605646 | ||||||||||||||||||||
Literature Links: |
SLC26A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC26A4 - solute carrier family 26 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000441.1 | 184 | Missense Mutation | GCC,TCC | A,S 61 | NP_000432.1 | |
XM_005250425.2 | 184 | Missense Mutation | GCC,TCC | A,S 61 | XP_005250482.1 | |
XM_006716025.3 | 184 | Missense Mutation | GCC,TCC | A,S 61 | XP_006716088.1 | |
XM_017012318.1 | 184 | Missense Mutation | GCC,TCC | A,S 61 | XP_016867807.1 |
SLC26A4-AS1 - SLC26A4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |