Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAAAATTTTTCCTTCACTCTGG[C/T]TCTTCCAGCCAGGCCCATGGTGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607905 MIM: 609214 | ||||||||||||||||||||
Literature Links: |
ALG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALG2 - ALG2, alpha-1,3/1,6-mannosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033087.3 | 1244 | Missense Mutation | ACC,GCC | T,A 392 | NP_149078.1 |
SEC61B - Sec61 translocon beta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |