Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAAAAAATCCCCCCATCCCCAA[C/T]CGTCTTTCACCAATACAAGAAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159558 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLLT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLLT3 - MLLT3, super elongation complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286691.1 | 3035 | UTR 3 | NP_001273620.1 | |||
NM_004529.3 | 3035 | UTR 3 | NP_004520.2 | |||
XM_017014726.1 | 3035 | UTR 3 | XP_016870215.1 | |||
XM_017014727.1 | 3035 | UTR 3 | XP_016870216.1 |