Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAAAAAATCCTAACCATTTTTG[C/T]TTCTTAAGTATTGTAAATACTGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615211 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UHRF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UHRF2 - ubiquitin like with PHD and ring finger domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152896.2 | Intron | NP_690856.1 | ||||
XM_011517704.2 | Intron | XP_011516006.1 | ||||
XM_011517705.2 | Intron | XP_011516007.1 | ||||
XM_017014253.1 | Intron | XP_016869742.1 | ||||
XM_017014254.1 | Intron | XP_016869743.1 |