Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTTGCTGTCTTCATTGCTCTC[A/C]CCGTTATGTTGTGGCTGATTACCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603620 MIM: 602348 | ||||||||||||||||||||
Literature Links: |
PSIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSIP1 - PC4 and SFRS1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128217.2 | 1826 | Silent Mutation | NP_001121689.1 | |||
NM_001317898.1 | 1826 | Intron | NP_001304827.1 | |||
NM_001317900.1 | 1826 | Intron | NP_001304829.1 | |||
NM_021144.3 | 1826 | Intron | NP_066967.3 | |||
NM_033222.4 | 1826 | Silent Mutation | NP_150091.2 |
SNAPC3 - small nuclear RNA activating complex polypeptide 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039697.1 | 1826 | Intron | NP_001034786.1 | |||
XM_017015056.1 | 1826 | Intron | XP_016870545.1 | |||
XM_017015057.1 | 1826 | Intron | XP_016870546.1 |