Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCCTCTCCTTTCCCTTTGAGAC[C/G]GTGAAGAGAAAGATGCAGGTGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300641 | ||||||||||||||||||||
Literature Links: |
SLC25A43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC25A43 - solute carrier family 25 member 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145305.2 | 781 | Silent Mutation | ACC,ACG | T,T 224 | NP_660348.2 | |
XM_017029340.1 | 781 | Silent Mutation | ACC,ACG | T,T 224 | XP_016884829.1 | |
XM_017029341.1 | 781 | Silent Mutation | ACC,ACG | T,T 224 | XP_016884830.1 |