Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGATTAAATCATTCAAGGCAA[A/C]AAAAGAGAAGGTTTAAAGTGGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCNT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCNT1 - glucosaminyl (N-acetyl) transferase 1, core 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097633.1 | Intron | NP_001091102.1 | ||||
NM_001097634.1 | Intron | NP_001091103.1 | ||||
NM_001097635.1 | Intron | NP_001091104.1 | ||||
NM_001097636.1 | Intron | NP_001091105.1 | ||||
NM_001490.4 | Intron | NP_001481.2 | ||||
XM_017014620.1 | Intron | XP_016870109.1 | ||||
XM_017014621.1 | Intron | XP_016870110.1 |