Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTGCACTGGCCGTAGCAGAAGC[C/G]GTTGAGGATGGTGCGGCTCCGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608832 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GREM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GREM2 - gremlin 2, DAN family BMP antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022469.3 | 563 | Missense Mutation | CGC,GGC | R,G 95 | NP_071914.3 | |
XM_005273226.3 | 563 | Missense Mutation | CGC,GGC | R,G 113 | XP_005273283.1 | |
XM_011544249.2 | 563 | Missense Mutation | CGC,GGC | R,G 95 | XP_011542551.1 |