Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCAGGCACACCCGGTTCCCAAG[A/G]ACGCCCGGATCACCCACTCCTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPN9 - calpain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319676.1 | 149 | Missense Mutation | AAC,GAC | N,D 18 | NP_001306605.1 | |
NM_006615.2 | 149 | Missense Mutation | AAC,GAC | N,D 18 | NP_006606.1 | |
NM_016452.2 | 149 | Missense Mutation | AAC,GAC | N,D 18 | NP_057536.1 | |
XM_011544017.1 | 149 | Missense Mutation | AAC,GAC | N,D 18 | XP_011542319.1 | |
XM_011544018.1 | 149 | Missense Mutation | AAC,GAC | N,D 18 | XP_011542320.1 | |
XM_011544019.2 | 149 | Missense Mutation | AAC,GAC | N,D 18 | XP_011542321.1 | |
XM_011544020.1 | 149 | Intron | XP_011542322.1 | |||
XM_017000098.1 | 149 | Missense Mutation | AAC,GAC | N,D 18 | XP_016855587.1 | |
XM_017000099.1 | 149 | Intron | XP_016855588.1 |