Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCATGACATTCATAAGGTTTC[C/T]TGCCAGAATGAATTTTCTGATGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608640 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF461 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF461 - zinc finger protein 461 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297623.2 | 1436 | Missense Mutation | AAG,AGG | K,R 387 | NP_001284552.1 | |
NM_001322821.1 | 1436 | Missense Mutation | AAG,AGG | K,R 404 | NP_001309750.1 | |
NM_001322823.1 | 1436 | Missense Mutation | AAG,AGG | K,R 379 | NP_001309752.1 | |
NM_001322825.1 | 1436 | Missense Mutation | AAG,AGG | K,R 379 | NP_001309754.1 | |
NM_001322826.1 | 1436 | Missense Mutation | AAG,AGG | K,R 356 | NP_001309755.1 | |
NM_001322827.1 | 1436 | Missense Mutation | AAG,AGG | K,R 356 | NP_001309756.1 | |
NM_001322828.1 | 1436 | Missense Mutation | AAG,AGG | K,R 322 | NP_001309757.1 | |
NM_153257.4 | 1436 | Missense Mutation | AAG,AGG | K,R 410 | NP_694989.2 | |
XM_011527486.2 | 1436 | Missense Mutation | AAG,AGG | K,R 283 | XP_011525788.1 | |
XM_017027470.1 | 1436 | Missense Mutation | AAG,AGG | K,R 366 | XP_016882959.1 |