Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATCTGAAAGTAGAGATGATGGC[A/C]GGGGGGACTCAGATCACACCCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613714 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376575 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105376575 - uncharacterized LOC105376575 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCR3LG1 - natural killer cell cytotoxicity receptor 3 ligand 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202439.2 | 1910 | Silent Mutation | GCA,GCC | A,A 32 | NP_001189368.1 | |
XM_011520072.2 | 1910 | Silent Mutation | GCA,GCC | A,A 3 | XP_011518374.1 | |
XM_011520073.2 | 1910 | Silent Mutation | GCA,GCC | A,A 3 | XP_011518375.1 | |
XM_011520074.2 | 1910 | Silent Mutation | GCA,GCC | A,A 3 | XP_011518376.1 | |
XM_011520075.2 | 1910 | Silent Mutation | GCA,GCC | A,A 3 | XP_011518377.1 |