Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGCTTCTACTGTCTTGCCATC[C/T]GGACAAACCAGCACGCTGGTCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IDH1 - isocitrate dehydrogenase (NADP(+)) 1, cytosolic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282386.1 | 1257 | Intron | NP_001269315.1 | |||
NM_001282387.1 | 1257 | Silent Mutation | CCA,CCG | P,P 298 | NP_001269316.1 | |
NM_005896.3 | 1257 | Intron | NP_005887.2 |