Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154550 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM219B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM219B - family with sequence similarity 219 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321920.1 | 1826 | Intron | NP_001308849.1 | |||
NM_001321921.1 | 1826 | Intron | NP_001308850.1 | |||
NM_001321922.1 | 1826 | Intron | NP_001308851.1 | |||
NM_001321923.1 | 1826 | Intron | NP_001308852.1 | |||
NM_020447.4 | 1826 | Intron | NP_065180.1 | |||
XM_017022433.1 | 1826 | Intron | XP_016877922.1 |
MPI - mannose phosphate isomerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289155.1 | 1826 | UTR 3 | NP_001276084.1 | |||
NM_001289156.1 | 1826 | UTR 3 | NP_001276085.1 | |||
NM_001289157.1 | 1826 | UTR 3 | NP_001276086.1 | |||
NM_002435.2 | 1826 | UTR 3 | NP_002426.1 | |||
XM_011521592.1 | 1826 | Intron | XP_011519894.1 | |||
XM_011521593.2 | 1826 | UTR 3 | XP_011519895.1 | |||
XM_017022207.1 | 1826 | Intron | XP_016877696.1 | |||
XM_017022208.1 | 1826 | Intron | XP_016877697.1 | |||
XM_017022209.1 | 1826 | Intron | XP_016877698.1 |
Set Membership: |
HapMap |