Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614666 MIM: 610998 MIM: 611118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC78 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC78 - coiled-coil domain containing 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031737.2 | 1289 | Missense Mutation | ACC,GCC | T,A 330 | NP_001026907.2 | |
XM_006720838.1 | 1289 | Missense Mutation | ACC,GCC | T,A 404 | XP_006720901.1 | |
XM_006720843.3 | 1289 | Missense Mutation | ACC,GCC | T,A 330 | XP_006720906.1 | |
XM_011522356.1 | 1289 | Missense Mutation | ACC,GCC | T,A 479 | XP_011520658.1 | |
XM_011522357.1 | 1289 | Missense Mutation | ACC,GCC | T,A 475 | XP_011520659.1 | |
XM_011522358.2 | 1289 | Missense Mutation | ACC,GCC | T,A 479 | XP_011520660.1 | |
XM_011522359.1 | 1289 | Missense Mutation | ACC,GCC | T,A 468 | XP_011520661.1 | |
XM_011522360.1 | 1289 | Missense Mutation | ACC,GCC | T,A 464 | XP_011520662.1 | |
XM_011522361.1 | 1289 | Missense Mutation | ACC,GCC | T,A 479 | XP_011520663.1 | |
XM_011522362.1 | 1289 | Missense Mutation | ACC,GCC | T,A 479 | XP_011520664.1 | |
XM_011522363.1 | 1289 | Missense Mutation | ACC,GCC | T,A 479 | XP_011520665.1 | |
XM_011522364.1 | 1289 | Missense Mutation | ACC,GCC | T,A 479 | XP_011520666.1 | |
XM_011522365.1 | 1289 | Missense Mutation | ACC,GCC | T,A 408 | XP_011520667.1 | |
XM_011522366.1 | 1289 | Missense Mutation | ACC,GCC | T,A 405 | XP_011520668.1 | |
XM_011522367.1 | 1289 | Missense Mutation | ACC,GCC | T,A 352 | XP_011520669.1 | |
XM_011522368.1 | 1289 | Missense Mutation | ACC,GCC | T,A 348 | XP_011520670.1 | |
XM_011522369.1 | 1289 | Missense Mutation | ACC,GCC | T,A 334 | XP_011520671.1 | |
XM_011522370.1 | 1289 | Missense Mutation | ACC,GCC | T,A 278 | XP_011520672.1 | |
XM_011522371.2 | 1289 | Missense Mutation | ACC,GCC | T,A 183 | XP_011520673.1 | |
XM_017022929.1 | 1289 | Missense Mutation | ACC,GCC | T,A 479 | XP_016878418.1 | |
XM_017022930.1 | 1289 | Missense Mutation | ACC,GCC | T,A 179 | XP_016878419.1 | |
XM_017022931.1 | 1289 | Intron | XP_016878420.1 | |||
XM_017022932.1 | 1289 | Missense Mutation | ACC,GCC | T,A 89 | XP_016878421.1 |
FAM173A - family with sequence similarity 173 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAGHL - hydroxyacylglutathione hydrolase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METRN - meteorin, glial cell differentiation regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NARFL - nuclear prelamin A recognition factor like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |