Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGAGAATTGACGAAGAGTTGAC[G/T]GGAAAATCCAGAAAATCTCAGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 170285 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP85 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUP85 - nucleoporin 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303276.1 | 577 | Silent Mutation | ACG,ACT | T,T 44 | NP_001290205.1 | |
NM_024844.4 | 577 | Silent Mutation | ACG,ACT | T,T 90 | NP_079120.1 | |
XM_005257690.2 | 577 | Silent Mutation | ACG,ACT | T,T 90 | XP_005257747.1 | |
XM_005257692.3 | 577 | UTR 5 | XP_005257749.1 | |||
XM_006722094.1 | 577 | Silent Mutation | ACG,ACT | T,T 90 | XP_006722157.1 | |
XM_011525267.1 | 577 | UTR 5 | XP_011523569.1 | |||
XM_011525268.1 | 577 | Silent Mutation | ACG,ACT | T,T 90 | XP_011523570.1 | |
XM_017025108.1 | 577 | Silent Mutation | ACG,ACT | T,T 90 | XP_016880597.1 | |
XM_017025109.1 | 577 | Silent Mutation | ACG,ACT | T,T 44 | XP_016880598.1 | |
XM_017025110.1 | 577 | UTR 5 | XP_016880599.1 |