Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCCCAGGTGCTGGGACAGGAC[A/G]ATGAGGTTGACCAGCGTCTCTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606919 MIM: 606942 MIM: 602880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CERS1 - ceramide synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COPE - coatomer protein complex subunit epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007263.3 | 800 | Silent Mutation | ATC,ATT | I,I 257 | NP_009194.2 | |
NM_199442.1 | 800 | Silent Mutation | ATC,ATT | I,I 206 | NP_955474.1 | |
NM_199444.1 | 800 | Silent Mutation | ATC,ATT | I,I 205 | NP_955476.1 | |
XM_011527655.1 | 800 | Silent Mutation | ATC,ATT | I,I 280 | XP_011525957.1 | |
XM_011527656.1 | 800 | Silent Mutation | ATC,ATT | I,I 228 | XP_011525958.1 |
GDF1 - growth differentiation factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |