Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGGGAGTGGACACCTGCAGTCT[C/T]TGCAGCGGCTGGTGAGTGTGTGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 120420 | ||||||||||||||||||||
Literature Links: |
CSF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSF1 - colony stimulating factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000757.5 | 564 | Silent Mutation | CTG,TTG | L,L 51 | NP_000748.3 | |
NM_172210.2 | 564 | Silent Mutation | CTG,TTG | L,L 51 | NP_757349.1 | |
NM_172211.3 | 564 | Silent Mutation | CTG,TTG | L,L 51 | NP_757350.1 | |
NM_172212.2 | 564 | Silent Mutation | CTG,TTG | L,L 51 | NP_757351.1 | |
XM_017000369.1 | 564 | Silent Mutation | CTG,TTG | L,L 10 | XP_016855858.1 |