Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATATGGTCCTCTAGCATTGGGT[A/G]ATGTGTATGAAAAGGGGCAAAAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TBCCD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBCCD1 - TBCC domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134415.1 | 1368 | Missense Mutation | CAC,TAC | H,Y 423 | NP_001127887.1 | |
NM_001286749.1 | 1368 | Missense Mutation | CAC,TAC | H,Y 327 | NP_001273678.1 | |
NM_018138.4 | 1368 | Intron | NP_060608.1 |