Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCAGACTGCGAAAAGTAGCTGGA[A/G]CCGGAGCAGGGACAGAACCTGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPFFR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPFFR2 - neuropeptide FF receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144756.1 | 316 | Intron | NP_001138228.1 | |||
NM_004885.2 | 316 | Missense Mutation | AAC,AGC | N,S 73 | NP_004876.2 | |
NM_053036.2 | 316 | Intron | NP_444264.1 | |||
XM_011531554.2 | 316 | Intron | XP_011529856.1 |