Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGAATTCTCACCTTTATCAGTC[A/G]GCGAATACTGTAAGAAATTAAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 111740 | ||||||||||||||||||||
Literature Links: |
GYPB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GYPB - glycophorin B (MNS blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304382.1 | 317 | Nonsense Mutation | CGA,TGA | R,* 61 | NP_001291311.1 | |
NM_002100.5 | 317 | Nonsense Mutation | CGA,TGA | R,* 87 | NP_002091.3 | |
XM_011531903.2 | 317 | Nonsense Mutation | CGA,TGA | R,* 87 | XP_011530205.1 | |
XM_011531904.2 | 317 | Intron | XP_011530206.1 | |||
XM_011531905.2 | 317 | Nonsense Mutation | CGA,TGA | R,* 78 | XP_011530207.1 | |
XM_017008137.1 | 317 | Nonsense Mutation | CGA,TGA | R,* 61 | XP_016863626.1 |