Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTCTGCAGCTTGCGGCTTTCA[G/T]TCACCCTTGGATTCAGAGATAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616798 | ||||||||||||||||||||
Literature Links: |
ZFP28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFP28 - ZFP28 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308440.1 | 890 | Intron | NP_001295369.1 | |||
NM_020828.1 | 890 | Intron | NP_065879.1 | |||
XM_011526462.2 | 890 | Intron | XP_011524764.1 | |||
XM_011526463.2 | 890 | Missense Mutation | CAG,CAT | Q,H 21 | XP_011524765.2 |
ZNF471 - zinc finger protein 471 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |