Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTAGACTGAATAGATGAAGGAT[A/G]TGCTTTTATTCTTTGGGGAAGGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP26 - ubiquitin specific peptidase 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031907.1 | 1561 | Missense Mutation | CAT,TAT | H,Y 475 | NP_114113.1 | |
XM_017029892.1 | 1561 | Missense Mutation | CAT,TAT | H,Y 475 | XP_016885381.1 |