Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAAGGCTCAGGTACTTCAAAGT[A/G]TGGCAGGGCAGACGCTAACCGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604531 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCR2 - natural cytotoxicity triggering receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199509.1 | 178 | Missense Mutation | ATG,GTG | M,V 31 | NP_001186438.1 | |
NM_001199510.1 | 178 | Missense Mutation | ATG,GTG | M,V 31 | NP_001186439.1 | |
NM_004828.3 | 178 | Missense Mutation | ATG,GTG | M,V 31 | NP_004819.2 | |
XM_017011500.1 | 178 | Missense Mutation | ATG,GTG | M,V 39 | XP_016866989.1 |