Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGGCCGGTGGAGTGCCCCCCA[C/T]GTGCCCACCCTGGCACTGCCCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610207 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC4A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC4A9 - solute carrier family 4 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258426.1 | 323 | Silent Mutation | CAC,CAT | H,H 96 | NP_001245355.1 | |
NM_001258427.1 | 323 | Silent Mutation | CAC,CAT | H,H 96 | NP_001245356.1 | |
NM_001258428.1 | 323 | Silent Mutation | CAC,CAT | H,H 120 | NP_001245357.1 | |
NM_031467.2 | 323 | Silent Mutation | CAC,CAT | H,H 96 | NP_113655.2 | |
XM_005268521.3 | 323 | Silent Mutation | CAC,CAT | H,H 120 | XP_005268578.2 | |
XM_011537694.2 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_011535996.1 | |
XM_017009933.1 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_016865422.1 | |
XM_017009934.1 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_016865423.1 | |
XM_017009935.1 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_016865424.1 | |
XM_017009936.1 | 323 | Silent Mutation | CAC,CAT | H,H 120 | XP_016865425.1 | |
XM_017009937.1 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_016865426.1 | |
XM_017009938.1 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_016865427.1 | |
XM_017009939.1 | 323 | Silent Mutation | CAC,CAT | H,H 120 | XP_016865428.1 | |
XM_017009940.1 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_016865429.1 | |
XM_017009941.1 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_016865430.1 | |
XM_017009942.1 | 323 | Silent Mutation | CAC,CAT | H,H 144 | XP_016865431.1 | |
XM_017009943.1 | 323 | Silent Mutation | CAC,CAT | H,H 120 | XP_016865432.1 |
Set Membership: |
HapMap Validated |