Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGTTAAGAATGCTGATCTCACC[A/G]AAGTAGCTGCCATCGCTCAATACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123825 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNGA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CNGA1 - cyclic nucleotide gated channel alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000087.3 | 1953 | Silent Mutation | TTC,TTT | F,F 544 | NP_000078.2 | |
NM_001142564.1 | 1953 | Silent Mutation | TTC,TTT | F,F 613 | NP_001136036.1 | |
XM_005248049.4 | 1953 | Silent Mutation | TTC,TTT | F,F 619 | XP_005248106.2 | |
XM_011513623.2 | 1953 | Silent Mutation | TTC,TTT | F,F 544 | XP_011511925.1 | |
XM_017007712.1 | 1953 | Silent Mutation | TTC,TTT | F,F 544 | XP_016863201.1 |
LOC101927157 - uncharacterized LOC101927157 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |