Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGCCTACTACAGGGACGCTGA[G/A]GCCTACAGACACTCCCCAGGTAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 305371 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GATA1 - GATA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002049.3 | 326 | Silent Mutation | GAA,GAG | E,E 67 | NP_002040.1 | |
XM_011543897.2 | 326 | Silent Mutation | GAA,GAG | E,E 67 | XP_011542199.1 | |
XM_011543898.2 | 326 | Intron | XP_011542200.1 |