Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606750 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZBP1 - Z-DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160417.1 | Intron | NP_001153889.1 | ||||
NM_001160418.1 | Intron | NP_001153890.1 | ||||
NM_001160419.2 | Intron | NP_001153891.1 | ||||
NM_001323966.1 | Intron | NP_001310895.1 | ||||
NM_030776.2 | Intron | NP_110403.2 | ||||
XM_011529057.2 | Intron | XP_011527359.1 | ||||
XM_011529058.2 | Intron | XP_011527360.1 | ||||
XM_017028086.1 | Intron | XP_016883575.1 |
Set Membership: |
HapMap |