Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606433 | |||||||||||||||||||||||||||||||||||
Literature Links: |
UGT1A8 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
UGT1A8 - UDP glucuronosyltransferase family 1 member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019076.4 | 783 | Silent Mutation | ACA,ACG | T,T 240 | NP_061949.3 |
Set Membership: |
HapMap DME Validated Inventoried |