Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCEPR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCEPR - cervical carcinoma expressed PCNA regulatory lncRNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHYHIPL - phytanoyl-CoA 2-hydroxylase interacting protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143774.1 | Intron | NP_001137246.1 | ||||
NM_032439.3 | Intron | NP_115815.2 | ||||
XM_011540275.2 | Intron | XP_011538577.1 | ||||
XM_011540276.2 | Intron | XP_011538578.1 | ||||
XM_017016782.1 | Intron | XP_016872271.1 | ||||
XM_017016783.1 | Intron | XP_016872272.1 |