Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602842 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMNN PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GMNN - geminin, DNA replication inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251989.1 | 517 | Intron | NP_001238918.1 | |||
NM_001251990.1 | 517 | Intron | NP_001238919.1 | |||
NM_001251991.1 | 517 | Intron | NP_001238920.1 | |||
NM_015895.4 | 517 | Intron | NP_056979.1 | |||
XM_005249159.1 | 517 | Intron | XP_005249216.1 | |||
XM_011514651.2 | 517 | UTR 5 | XP_011512953.1 |